Learn More
Cayman Chemical NudIfloramIde 10mg
SDP
Catalog No. 502982336
Encompass_Preferred
An active metabolite of nicotinamide; an inhibitor of PARP (IC50 = 8 µM); plasma levels are increased in patients with chronic renal failure
Catalog No. 50-298-2336 Supplier Cayman Chemical Supplier No. 2050710 MG
An active metabolite of nicotinamide; an inhibitor of PARP (IC50 = 8 µM); plasma levels are increased in patients with chronic renal failure