Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nudifloramide-d3 | 1207384-48-2 | 98.5% | 155.17 g/mol | C7H5D3N2O2 | 5 MG
SDP

Catalog No. 5000263919
Encompass_Preferred

Nudifloramide-d3 is the deuterium-labeled isotopologue of nudifloramide (2PY), provided as a stable-isotope analytical standard intended for research and analytical applications. It is suitable for use in metabolomics and trace-quantification workflows and is reported to inhibit poly(ADP-ribose) polymerase (PARP-1) activity in vitro.

  • Deuterium-labeled internal standard for analytical assays.
  • Suitable for metabolomics and trace-level quantification.
  • Molecular formula C7H5D3N2O2; molecular weight 155.17 g/mol.
  • High purity (approximately 98.5%) for reliable results.
  • Supplied as a small pack suitable for analytical use (5 mg).

Catalog No. 50-002-63919 Supplier Medchemexpress LLC Supplier No. HY113432S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nudifloramide-d3 is the deuterium-labeled isotopologue of nudifloramide (2PY), provided as a stable-isotope analytical standard intended for research and analytical applications. It is suitable for use in metabolomics and trace-quantification workflows and is reported to inhibit poly(ADP-ribose) polymerase (PARP-1) activity in vitro.

  • Deuterium-labeled internal standard for analytical assays.
  • Suitable for metabolomics and trace-level quantification.
  • Molecular formula C7H5D3N2O2; molecular weight 155.17 g/mol.
  • High purity (approximately 98.5%) for reliable results.
  • Supplied as a small pack suitable for analytical use (5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.