Learn More
Medchemexpress LLC Nudifloramide-d3 | 1207384-48-2 | 99.4% | 155.17 | 1 MG
SDP
Catalog No. 5000436809
Encompass_Preferred
Nudifloramide-d3 (2PY-d3) is a deuterium labeled Nudifloramide. It is an end product of nicotinamide-adenine dinucleotide (NAD) degradation and significantly inhibits poly(ADP-ribose) polymerase (PARP-1) activity in vitro. Deuteration, the incorporation of stable heavy isotopes, allows it to be used as a tracer during drug development, affecting pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36809 Supplier Medchemexpress LLC Supplier No. HY113432S1MG
Nudifloramide-d3 (2PY-d3) is a deuterium labeled Nudifloramide. It is an end product of nicotinamide-adenine dinucleotide (NAD) degradation and significantly inhibits poly(ADP-ribose) polymerase (PARP-1) activity in vitro. Deuteration, the incorporation of stable heavy isotopes, allows it to be used as a tracer during drug development, affecting pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.