Learn More
Aviva Systems Biology Corp Nudt15 Antibody - N-terminal region (ARP48830_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48830_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nudt15. It was validated on Western Blot. . Alternate names: 6530403O17, A730068G11Rik, MTH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-381