Learn More
Aviva Systems Biology Corp Nudt16 Antibody - N-terminal region (ARP52936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nudt16. It was validated on Western Blot. . Alternate names: 2310041H06Rik, 2810047L04Rik, 2900006H04Rik, AI851783ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-789