Learn More
Aviva Systems Biology Corp NUDT16L1 Antibody - middle region (ARP48417_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48417_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT16L1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NUDT16L1. Alternate names: MGC11275, SDOSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-360