Learn More
Aviva Systems Biology Corp NUDT17 Antibody - middle region (ARP48509_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48509_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NUDT17. Alternate names: FLJ34433ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53558