Learn More
Aviva Systems Biology Corp NUDT18 Antibody - N-terminal region (ARP48921_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48921_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NUDT18. Alternate names: FLJ22494ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rat
Catalog No. 50-014-59960