Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Oxide, Red, Powder, Technical, Spectrum™ Chemical
SDP

Catalog No. 18602675 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-675 45 kg Amber Glass Bottle
18-602-672 500 g Amber Glass Bottle
18-602-673 2.5 kg Amber Glass Bottle
18-602-674 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-675 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F104045KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F1040, 1309-37-1, Fe2O3

Spectrum™ Chemical Ferric Oxide, Red, Powder, Technical , also known as iron(II) oxide, is one of the main oxides of iron. It is sometimes referred to as rust and also occurs naturally as the mineral magnetite. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

CAS Max % 100%
Physical Form Powder, Needles or Crystals
Quantity 45 kg
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Ferric Oxide, Red, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.