Learn More
Aviva Systems Biology Corp NUDT2 Antibody - middle region (ARP62697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT2. It was validated on Western Blot. . Alternate names: APAH1, MGC10404ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-746