Learn More
Aviva Systems Biology Corp NUDT21 Antibody - N-terminal region (ARP40695_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40695_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT21. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human NUDT21. Alternate names: CPSF5, CFIM25ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57591