Learn More
Aviva Systems Biology Corp NUDT22 Antibody - C-terminal region (ARP60482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT22. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NUDT22. Alternate names: MGC13045ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-059