Learn More
Aviva Systems Biology Corp NUDT6 Antibody - middle region (ARP62064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUDT6. It was validated on Western Blot. . Alternate names: ASFGF2, FGF-2, FGF-AS, FGF2AS, bFGF, gfg, gfg-1, GFG1, GFG-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-555