Learn More
ATCC NU-DUL-1; Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL2969
Encompass
NU-DUL-1 is a lymphocyte cell line that was isolated in 1985 from the peritoneal effusion of a 43-year-old, White male with lymphoma non-Burkitt's type. It can be used for immunology research.
Catalog No. 50-238-2201