Learn More
Aviva Systems Biology Corp NUFIP2 Antibody - N-terminal region (ARP51676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUFIP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NUFIP2. Alternate names: 182-FIP, 82-FIP, FIP-82, FLJ10976, KIAA1321, MGC117262, PIG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60582