Learn More
Med Vet International Nuflor Injection, 300mg/mL, 250mL, Florfenicol
Catalog No. 502827801
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Nuflor Injectable Solution (florfenicol) is a synthetic broad-spectrum antibiotic for IM and SQ use in cattle only. Indicated for treatment of bovine respiratory disease (BRD) associated with Mannheimia (Pasteurella) haemolytica, Pasteurella multocida, and Haemophilus somnus, and for the treatment of bovine interdigital phlegmon (foot rot, acute interdigital necrobacillosis, infectious pododermatitis) associated with Fusobacterium necrophorum and Bacteroides melaninogenicus. Also, it is indicated for the control of respiratory disease in cattle at high risk of developing BRD associated with Mannheimia (Pasteurella) haemolytica, Pasteurella multocida, and Haemophilus somnus.
Catalog No. 50-282-7801 Supplier Med Vet International Supplier No. RXNUFLOR250ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Nuflor Injectable Solution (florfenicol) is a synthetic broad-spectrum antibiotic for IM and SQ use in cattle only. Indicated for treatment of bovine respiratory disease (BRD) associated with Mannheimia (Pasteurella) haemolytica, Pasteurella multocida, and Haemophilus somnus, and for the treatment of bovine interdigital phlegmon (foot rot, acute interdigital necrobacillosis, infectious pododermatitis) associated with Fusobacterium necrophorum and Bacteroides melaninogenicus. Also, it is indicated for the control of respiratory disease in cattle at high risk of developing BRD associated with Mannheimia (Pasteurella) haemolytica, Pasteurella multocida, and Haemophilus somnus.