Learn More
World Precision Instrument Nugent Utility Forceps, 10mm, Angled Tip
SDP
Catalog No. NC0696847
Encompass
Nugent Utility Forceps are designed for a wide range of uses in various fields, including medical, dental, laboratory, and other applications. Forceps are handheld, hinged instruments used for grasping, holding, or manipulating objects or tissues during medical procedures, surgeries, or laboratory work. The tips of the forceps are angled or straight, and the tips are serrated for a firm grasp. The thin, narrow tips allow access in small areas. The ribbed handles give you a solid grip, letting you apply an appropriate amount of pressure with some tactile feedback. WPI Nugent Forceps are manufactured from surgical grade stainless steel and are suitable for sterilization via autoclave or chemical sterilants.
Catalog No. NC0696847 Supplier World Precision Instrument Supplier No. 504473
Nugent Utility Forceps are designed for a wide range of uses in various fields, including medical, dental, laboratory, and other applications. Forceps are handheld, hinged instruments used for grasping, holding, or manipulating objects or tissues during medical procedures, surgeries, or laboratory work. The tips of the forceps are angled or straight, and the tips are serrated for a firm grasp. The thin, narrow tips allow access in small areas. The ribbed handles give you a solid grip, letting you apply an appropriate amount of pressure with some tactile feedback. WPI Nugent Forceps are manufactured from surgical grade stainless steel and are suitable for sterilization via autoclave or chemical sterilants.