Learn More
International Crystal Nujol high purity mineral oil 60ml
Supplier:  International Crystal 00112475
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Nujol; Mineral Oil, 60mL, high Purity, for Preparation of Mull Samples in the Wavelength Range of 1370cm-1 to the Far Infrared, with Only One Absorption Band at 720Cm-1, Ideal for Preparing Mulls w/Real Crystal; IR Cards or Salt Crystals
Catalog No. NC9697623