Learn More
Enzo Life Sciences Nullscript (5mg). CAS: 300816-11-9
SDP
Supplier:  Enzo Life Sciences BMLGR3270005
Encompass_Preferred
Negative control compound for the histone deacetylase (HDAC) inhibitor scriptaid (BML-GR326). Alternative name: 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (4mg/ml) or ethanol (0.3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0983