Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4926MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: A73-B/D12. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Exponentially growing any cultured human cells. Tonsil or lymph node. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Colon carcinoma 174T cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 228kDa.Chromosome: 11q13. Gene Symbol: NUMA1.
Catalog No. 50-188-5024