Learn More
Grainger Young Bros. Stamp Works Hand Stamp Kit, Steel, 90 to 99, 1/4 in. Character Height, 1 ea.
Catalog No. 502786352
Encompass_Preferred
This heavy-duty hand stamp kit is designed for marking applications requiring double-digit numerical impressions. Constructed from durable steel, the set features a round face impression style and is suitable for materials with a maximum surface hardness of Rockwell C50.
  • Character height: 1/4 in.
  • Shank length: 3 in.
  • Shank size: 9/16 in.
  • Set includes: (10) stamps for numbers 90 to 99
  • Material: Steel
  • Origin: USA
Catalog No. 50-278-6352 Supplier Grainger Supplier No. 18G311
This heavy-duty hand stamp kit is designed for marking applications requiring double-digit numerical impressions. Constructed from durable steel, the set features a round face impression style and is suitable for materials with a maximum surface hardness of Rockwell C50.
  • Character height: 1/4 in.
  • Shank length: 3 in.
  • Shank size: 9/16 in.
  • Set includes: (10) stamps for numbers 90 to 99
  • Material: Steel
  • Origin: USA