Learn More
Electron Microscopy Sciences Numbered Grid Storage Box
SDP
Supplier:  Electron Microscopy Sciences 71138
Encompass_Preferred
The standard 100 capacity grid storage box with a unique number printed on the face and on one end. Eliminates the placement of the grid in the wrong box. Easy retrieval of grid box from storage. Complete with grid recording card.
Catalog No. 50-930-845