Learn More
Grainger Numbers And Letters Dash 3/4In H PK10
Catalog No. 502750384
Encompass_Preferred
The Brady Letter 802GT4 is a white vinyl label with a permanent acrylic adhesive that is designed for indoor use on smooth, flat surfaces like metal, glass, plastic, etc. It measures 4" x 6" and comes in a pack of 250 labels. The labels have a matte finish and feature a strong adhesive that adheres well to low surface energy plastics. They are resistant to UV light, chemicals, moisture, and abrasion. These vinyl asset tags can be used for barcoding, equipment ID, wire marking, and general information labeling indoors. They have an operating temperature range of -40°F to 176°F.
Catalog No. 50-275-0384 Supplier Grainger Supplier No. 14R064
The Brady Letter 802GT4 is a white vinyl label with a permanent acrylic adhesive that is designed for indoor use on smooth, flat surfaces like metal, glass, plastic, etc. It measures 4" x 6" and comes in a pack of 250 labels. The labels have a matte finish and feature a strong adhesive that adheres well to low surface energy plastics. They are resistant to UV light, chemicals, moisture, and abrasion. These vinyl asset tags can be used for barcoding, equipment ID, wire marking, and general information labeling indoors. They have an operating temperature range of -40°F to 176°F.