Learn More
Grainger Numbers And Letters Kit A Thru Z
Catalog No. 502750389
Encompass_Preferred
The Brady Number Label is a 2 inch character height label made of vinyl with a permanent acrylic adhesive. It has high visibility and is resistant to most industrial chemicals and solvents. The label comes in white with black print. It can be used for pipe marking, equipment ID, valve and instrument tagging, and other identification applications. The label meets ANSI standards for pipe marking.
Catalog No. 50-275-0389 Supplier Grainger Supplier No. 14R097
The Brady Number Label is a 2 inch character height label made of vinyl with a permanent acrylic adhesive. It has high visibility and is resistant to most industrial chemicals and solvents. The label comes in white with black print. It can be used for pipe marking, equipment ID, valve and instrument tagging, and other identification applications. The label meets ANSI standards for pipe marking.