Learn More
Medchemexpress LLC Numidargistat dihydrochloride | 98.5% | 360.04 g·mol^-1 | C11H24BCl2N3O5 | 100 MG
SDP
Catalog No. 5000247921
Encompass_Preferred
Numidargistat dihydrochloride is a research-grade small-molecule inhibitor of arginase used in preclinical immunology and oncology studies. It displays nanomolar activity against recombinant human arginase enzymes and is supplied as the dihydrochloride salt with manufacturer guidance for storage and batch-specific purity.
  • Potent inhibition of arginase 1 and 2 (IC50 ≈ 86 nM and 296 nM)
  • Orally bioavailable profile suitable for in vivo dosing in preclinical models
  • Suitable for in vitro biochemical and cell-based assays
  • Dihydrochloride salt form improves solubility and handling
  • Supplied with batch COA and storage recommendations for research use only
Catalog No. 50-002-47921 Supplier Medchemexpress LLC Supplier No. HY101979A100MG
Numidargistat dihydrochloride is a research-grade small-molecule inhibitor of arginase used in preclinical immunology and oncology studies. It displays nanomolar activity against recombinant human arginase enzymes and is supplied as the dihydrochloride salt with manufacturer guidance for storage and batch-specific purity.
  • Potent inhibition of arginase 1 and 2 (IC50 ≈ 86 nM and 296 nM)
  • Orally bioavailable profile suitable for in vivo dosing in preclinical models
  • Suitable for in vitro biochemical and cell-based assays
  • Dihydrochloride salt form improves solubility and handling
  • Supplied with batch COA and storage recommendations for research use only