Learn More
Medchemexpress LLC Numidargistat dihydrochloride | 99.8% | 360.04 g/mol | C11H24BCl2N3O5 | 50 MG
SDP
Catalog No. 5000246533
Encompass_Preferred
Numidargistat dihydrochloride is a small-molecule arginase inhibitor supplied for research use in immuno-oncology and metabolic enzyme studies. It shows low-nanomolar inhibition of human arginase isoforms and has been used to modulate the tumor microenvironment by reducing myeloid cell-mediated immune suppression. The dihydrochloride salt is provided to improve handling and solubility for in vitro and in vivo experiments.
  • Potent arginase inhibition with low-nanomolar activity.
  • Used to modulate tumor immune microenvironment in research models.
  • Dihydrochloride salt form supports improved solubility and handling.
  • High reported purity for research applications (reported ≈99.8%).
  • Available in small research quantities suitable for preclinical studies.
Catalog No. 50-002-46533 Supplier Medchemexpress LLC Supplier No. HY101979A50MG
Numidargistat dihydrochloride is a small-molecule arginase inhibitor supplied for research use in immuno-oncology and metabolic enzyme studies. It shows low-nanomolar inhibition of human arginase isoforms and has been used to modulate the tumor microenvironment by reducing myeloid cell-mediated immune suppression. The dihydrochloride salt is provided to improve handling and solubility for in vitro and in vivo experiments.
  • Potent arginase inhibition with low-nanomolar activity.
  • Used to modulate tumor immune microenvironment in research models.
  • Dihydrochloride salt form supports improved solubility and handling.
  • High reported purity for research applications (reported ≈99.8%).
  • Available in small research quantities suitable for preclinical studies.