Learn More
Thermo Scientific™ Nunc™ 1536-Well Microplates Plates
Catalog No. 12565452 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
White
Catalog No. Color
12-565-452 White
1 options
Catalog No. 12-565-452 Supplier Thermo Scientific™ Supplier No. 253607
Screen four times as many samples in a single plate with Thermo Scientifi™ Nunc™ 1536-Well Microplates, which are ideal for high-throughput screening.
Maximize high-throughput screening efficiency by using Thermo Scientific™ Nunc™ 1536-Well Microplates, built according to ANSI standards and with the highest quality.
  • Low plate height is optimized for stacking
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working well volume, 1 to 10μL
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid Without Lid
Array 32 x 48
Packaging 90 Cases
Product Line Nunc
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 1 to 10 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 90
No. per Pack 30
Quantity 90/Cs.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.