Learn More
Thermo Scientific™ Nunc™ MicroWell™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom, Optical Polymer Base Microplate
Catalog No. 12565905 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 20
Catalog No. Quantity
12-565-905 Case of 20
1 options
Catalog No. 12-565-905 Supplier Thermo Scientific™ Supplier No. 152029
Add to Cart
Add to Cart
Color:Black, Clear Bottom; Lid:With Lid; Type: Optical Polymer Base
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 384-Well Optical-Bottom Plates with Polymer or Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Poly- D-Lysine and Collagen coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
  • Cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
  • Polystyrene upper structure is available with either a #1.5 cover glass or polystyrene base
  • Customized bar coding is available
  • Aseptically Manufactured
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Volume (Metric) Well 120 μL
Type Optical Polymer Base
Quantity Case of 20
No. per Pack 5
No. per Case 20
Color Black
Surface Treatment Poly-D-Lysine
Sterility Aseptically Manufactured
No. of Wells 384
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Array 16 x 24
Product Line Nunc
Packaging 10 Pk., 30 Cases
Barcode Custom Barcoding Available
Well Shape Flat
Show More Show Less
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.