Learn More
Thermo Scientific™ 384 Well Black Plate, Non-Treated Surface, Non-Sterile, Pack of 25
Catalog No. 1256854 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 100
Catalog No. Quantity
12-568-54 Case of 100
1 options
Catalog No. 12-568-54 Supplier Thermo Scientific™ Supplier No. 262260
Add to Cart
Add to Cart
Thermo Scientific™ Nunc™ black microplates are ideal for fluorescence measurements, with minimum back-scattered light and background fluorescence.
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc™ 384-Well Polystyrene Black and White Microplates.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid Without Lid
Culture Area 0.084 cm2/well
Array 16 x 24
Packaging 25/Pk., 100/Cs.
Product Line Nunc
Color Black
Product Type Microplate
Skirt Style Low Flange Pinchbar
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 100
No. per Pack 25
Quantity Case of 100
Show More Show Less
Do you offer Nunc Microplate Lids for the Nunc Black or White 384-Well Microplates?
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.
Why do you offer Nunc 384-Well Black or White Microplates when the cells cannot be observed using an inverted light microscope?
We offer Nunc Black or White 384-Well microplates that are intended for use in conjunction with 384-well microplate luminometers and fluorometers that have excitation and detection systems located above the plate. Due to the opaque quality of these microplates, all detection systems must rely on either luminescent or fluorescent reagents, such as viability stains that report cell activity and membrane integrity.
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.