Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ IVF Multi Dish, 4-well

Catalog No. 12566300
Change view
Click to view available options
Surface Treatment:
Non-treated
Nunclon Delta
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
12-566-300 Nunclon Delta
12-566-350 Non-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-566-300 Supplier Thermo Scientific™ Supplier No. 144444
Save 50% on Cell Culture Plastics
Use promo code NUNC50
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform applications which yield consistently reliable, high-quality results during vitrification.

Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.

Innovative features for high quality and ease-of-use:

  • Ensures proper well orientation and prevents cross contamination with notched lid and dish
  • Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
  • The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
  • The non-treated hydrophobic surface facilitates microdrop embryo culture
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
Description Nunclon Delta-treated IVF Dish, 4-well
Culture Area 1.9 cm2/well
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Nunclon Delta
Well Count 4
No. per Case 120
No. per Pack 4
Length (Metric) 66 mm
Width (Metric) 66 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.