Learn More
Description
Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.
Innovative features for high quality and ease-of-use:
- Ensures proper well orientation and prevents cross contamination with notched lid and dish
- Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
- The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
- The non-treated hydrophobic surface facilitates microdrop embryo culture
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
Specifications
| Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
| Description | Non-treated IVF Dish, 4-well |
| Culture Area | 1.9 cm2/well |
| Material | Polystyrene (USP Class VI) |
| Sterility | Sterile |
| Surface Treatment | Untreated |
| Well Count | 4 |
| No. per Case | 120 |
| No. per Pack | 4 |
| Length (Metric) | 66 mm |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.