Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates
GSA_VA

Catalog No. 12565553 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
12-565-553 Sterile
12-565-552 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-565-553 Supplier Thermo Scientific™ Supplier No. 278606
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clone and replicate assays with these untreated polystyrene plates, suited for agglutination assays and sample collection.

Store samples and perform agglutination assays with Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates, featuring transparent round bottom deep wells that hold up to 900μL per well.

  • Alphanumeric grid provides quick and easy sample identification.
  • Optimized for robotics and liquid handling systems
  • Round-bottom wells for optimal sample recovery
  • Untreated surface
  • Sealable with well caps and sealing tape
  • Customized bar coding is available

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Packaging 32 Cases, 4 Pk.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 900 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 32
No. per Pack 4
Quantity 32/Cs.
Dimensions (L x W x H) 128 x 86 x 41 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.