Learn More
Thermo Scientific™ 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Catalog No. 12567209 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 50
Catalog No. Quantity
12-567-209 Case of 50
1 options
Catalog No. 12-567-209 Supplier Thermo Scientific™ Supplier No. 277143
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart
Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems.
Thermo Scientific Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well conical bottom (V) microplates are ideal for sample recovery.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
No. per Case 50
No. per Pack 1
Quantity Case of 50
Show More Show Less
Is the Magnetic Stand-96 (Cat. No. AM10027) compatible with 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 (Cat. No. 277143)?
Yes. The Magnetic Stand-96 (Cat. No. AM10027) is compatible with 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 (Cat. No. 277143).