Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
GSA_VA
Catalog No. 12565215
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
No. per Case:
160/Cs.
180/Cs.
50/Cs.
80/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Lid No. per Case
12-565-215 Sterile Without Lid 50/Cs.
12-565-216 Non-sterile Without Lid 180/Cs.
12-565-481 Sterile With Lid 80/Cs.
12-565-487 Sterile Without Lid 80/Cs.
12-565-460 Sterile With Lid 160/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 12-565-215 Supplier Thermo Scientific™ Supplier No. 249662
Save 50% on Cell Culture Plastics
Use promo code NUNC50
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
Features:
  • Conical bottom (V) microplates are ideal for sample recovery.
    • Applications include antibiotic screens, serological tests, storing and screening compounds, DNA libraries, and titration of solutions.
Warranty: 90 days
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 50 Cases
Color Clear
Product Type Microplate
Skirt Style Low Flange Pinchbar
Material Polystyrene
Volume (Metric) Well 300 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
No. per Case 50/Cs.
No. per Pack 1 Ea.
Quantity 1 Ea.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.