Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Catalog No. 12565481 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
No. per Case:
160/Cs.
180/Cs.
50/Cs.
80/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Lid No. per Case
12-565-481 Sterile With Lid 80/Cs.
12-565-216 Non-sterile Without Lid 180/Cs.
12-565-215 Sterile Without Lid 50/Cs.
12-565-487 Sterile Without Lid 80/Cs.
12-565-460 Sterile With Lid 160/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-565-481 Supplier Thermo Scientific™ Supplier No. 249935
  • $642.33
    $523.65 / Case of 80
    Save $118.68
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
Features:
  • Conical bottom (V) microplates are ideal for sample recovery.
    • Applications include antibiotic screens, serological tests, storing and screening compounds, DNA libraries, and titration of solutions.
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid With Lid
Array 8 x 12
Packaging 80 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
No. per Case 80/Cs.
No. per Pack 10 Pk.
Quantity 80/Cs.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.