Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Catalog No. 12565212
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
No. per Pack:
1
10
135
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Lid No. per Pack
12-565-212 Sterile Without Lid 1
12-565-213 Non-sterile Without Lid 135
12-565-214 Non-sterile Without Lid 10
12-565-331 Sterile With Lid 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-565-212 Supplier Thermo Scientific™ Supplier No. 262162
Save 50% on Cell Culture Plastics
Use promo code NUNC50
View all promotions
Add to Cart
Edge
Add to Cart
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
  • Clear polystyrene has excellent optical properties ideal for optical measurements
  • Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
  • Customized bar coding is available
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 1 Pk.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 50
No. per Pack 1
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.