Learn More
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Catalog No. 09552182
Change view
Click to view available options
Capacity (Metric):
1 mL
1.8 mL
4.5 mL
Closure Color:
Blue
Green
Orange
Purple
Red
Yellow
Bottom Shape:
Conical (Starfoot)
Round (Starfoot)
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color Bottom Shape
09-552-182 4.5 mL Red Round (Starfoot)
09-552-152 4.5 mL Orange Round (Starfoot)
09-552-175 4.5 mL Yellow Round (Starfoot)
09-552-254 4.5 mL Green Round (Starfoot)
09-552-123 4.5 mL Blue Round (Starfoot)
09-552-127 4.5 mL Purple Round (Starfoot)
09-552-137 1 mL Red Conical (Starfoot)
09-552-165 1 mL Orange Conical (Starfoot)
09-552-138 1 mL Yellow Conical (Starfoot)
09-552-268 1 mL Green Conical (Starfoot)
09-552-241 1 mL Blue Conical (Starfoot)
09-552-216 1 mL Purple Conical (Starfoot)
09-552-219 1.8 mL Red Round (Starfoot)
09-552-253 1.8 mL Orange Round (Starfoot)
09-552-220 1.8 mL Yellow Round (Starfoot)
09-552-150 1.8 mL Green Round (Starfoot)
09-552-196 1.8 mL Blue Round (Starfoot)
09-552-197 1.8 mL Purple Round (Starfoot)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 09-552-182 Supplier Thermo Scientific™ Supplier No. 337516RED
Only null left
Add to Cart
Add to Cart
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Thermo Scientific™ Nunc Cryotubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage applications.
  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryogenic Storage Tube
Closure Color Red
Autoclavable Not Autoclavable
Capacity (Metric) 4.5 mL
Bottom Shape Round (Starfoot)
Material Polypropylene
Thread Style External
Graduations Yes
Style Writing Surface, A-R Base
Sterility Sterile
Graduated Yes