Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Cell-Culture Treated Multidishes

Catalog No. 1483211 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Wells:
4
6
12
24
48
No. per Case:
120/Cs.
75/Cs.
85/Cs.
No. per Pack:
1 Ea.
4 Pk.
5 Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells No. per Case No. per Pack
14-832-11 6 75/Cs. 1 Ea.
12-565-163 24 75/Cs. 1 Ea.
12-565-321 12 75/Cs. 1 Ea.
12-565-322 48 75/Cs. 1 Ea.
12-565-72 4 120/Cs. 4 Pk.
14-832-10 6 85/Cs. 5 Pk.
12-565-164 24 85/Cs. 5 Pk.
12-565-165 48 85/Cs. 5 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-832-11 Supplier Thermo Scientific™ Supplier No. 140675
  • $471.76
    $319.65 / Case of 75

    Save $152.11

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nunc Cell-Culture Treated Multidishes receive the proprietary Nunclon Delta surface treatment that promotes maximum adhesion for a broad range of cell types.

Thermo Scientific Nunc Cell-Culture Treated Multidishes receive the Nunclon Delta surface treatment. This proprietary surface modification promotes maximum adhesion for a broad range of cell types and ensures optimum performance.

Nunclon Delta surface treatment ensures optimum performance for cell adhesion.

  • Flat well bottom and stackable
  • Raised well rims lower risk of cross contamination
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Excellent optical quality
  • Alphanumerical codes for well identification

Round well styles

  • Vented lids for gas exchange
  • Recessed areas around wells act as a water reservoir to reduce evaporation from wells
  • Lids with notched corners for easy orientation

Rectangular well styles

  • Large culture area
  • Accept standard glass cover slips: 24 x 60 mm for 4-well plate and 22 x 22 mm for 8-well plate

Additional information

24-well plates are 17 mm deep; 4-well plate is 10 mm deep

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance With certificate of traceability and quality (one certificate per case).
Format SBS Standard
Culture Area 9.6 sq. cm
Array 2 x 3
Packaging Individually Packed
Color Clear
Product Type Cell Culture Dish
Material Polystyrene
Volume (Metric) Well 3 mL (Well)
No. of Wells 6
Volume (Metric) Working 3 mL
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
Includes Lid
No. per Case 75/Cs.
No. per Pack 1 Ea.
Quantity 1 Ea.
Length (Metric) 127.76 mm
Width (Metric) 85.46 mm
Show More Show Less
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.