Description
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
- Plug a media bag or tubing set into any cell factory
- Cleaning processes and sterility validations are minimized
- Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
- Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
Specifications
Specifications
| Description | Tubing Set with Male MPC connector |
| For Use With (Equipment) | Cell Factory Systems (All) |
| No. per Pack | 1 |
| Product Type | Tubing Set, Male MPC |
| Sterility | Sterile |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.