Learn More
Cole-Parmer Thermo Scientific Nunc 378220 Chipboard Storage Box, 6/cs
Catalog No. NC9931300 Shop All Cole-Parmer Products
Encompass
Each 5 1/8"L X 5 1/8"W X 2"D box holds up to 81 cryogenic vials (1.0 to 1.8 mL).
Catalog No. NC9931300 Supplier Cole-Parmer Supplier No. 0375900
Each 5 1/8"L X 5 1/8"W X 2"D box holds up to 81 cryogenic vials (1.0 to 1.8 mL).