Learn More
Thermo Scientific™ Nunc™ CryoTube™ Colored Caps
Catalog No. 12565169
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Red
Yellow
Sterility:
Non-sterile
Sterile
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility
12-565-169 Red Sterile
12-565-168 Blue Sterile
12-565-167 Yellow Sterile
12-565-170 Assorted Sterile
12-567-00 Clear Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Do not use CryoTubes in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ Tubing (Cat. No. 343958). Improper use may cause liquefied nitrogen to be trapped inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nunc Cryopreservation Manual when handling and disposing of the CryoTubes.
Catalog No. 12-565-169 Supplier Thermo Scientific™ Supplier No. 336081
Only null left
Add to Cart
Add to Cart
Do not use CryoTubes in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ Tubing (Cat. No. 343958). Improper use may cause liquefied nitrogen to be trapped inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nunc Cryopreservation Manual when handling and disposing of the CryoTubes.
Color-code your sample vials with replacement polypropylene screw caps featuring a silicone gasket to provide the best seal for internally threaded cryotube vials.
Color-code your sample vials. Thermo Scientific™ Nunc™ CryoTube™ Colored Caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal. Nunc CryoTubes are the most widely used cryo vials in the world, stemming from the first cryo vials manufactured in the early 1960s. The well-proven design, the high certification level and the large portfolio of vials provide researchers with a choice of the vial best suited for their application and peace of mind that the sample can be retrieved viable and unharmed.
  • These replacement polypropylene screw caps have a silicone gasket to provide the best seal
  • Available nonsterile or irradiated
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material Polypropylene
Sterility Sterile
Product Line Nunc
Quantity 250/Pk., 2500/Cs.
Product Type Cap