Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

Catalog No. 12565671 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Culture Area:
145 cm2
21.5 cm2
56.7 cm2
8.8 cm2
Height (Metric):
13 mm
16 mm
17 mm
21 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Culture Area Height (Metric)
12-565-671 35 mm 8.8 cm2 13 mm
12-565-670 60 mm 21.5 cm2 16 mm
12-565-668 100 mm 56.7 cm2 21 mm
12-565-669 100 mm 56.7 cm2 17 mm
12-565-667 150 mm 145 cm2 21 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-565-671 Supplier Thermo Scientific™ Supplier No. 150470
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Culture Area 8.8 cm2
Disposable Single-use
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Nunclon Supra
Type Cell Culture Dish
No. per Case 500 Dishes
No. per Pack 10 Dishes
Height (Metric) 13 mm
Diameter (Metric) 35 mm
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.