Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface

Catalog No. 12565662
Change view
Click to view available options
Culture Area:
175 cm²
25 cm²
75 cm²
Closure Type:
Filter
Solid
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Closure Type
12-565-662 175 cm² Filter
12-565-666 25 cm² Filter
12-565-665 25 cm² Solid
12-565-664 75 cm² Filter
12-565-663 75 cm² Solid
12-565-661 175 cm² Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-565-662 Supplier Thermo Scientific™ Supplier No. 156380
Only null left
Add to Cart
Edge
Add to Cart

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Description Nunc EasYFlask, 175 cm²
No. per Case 30 Flasks
No. per Pack 5 Flasks
Closure Type Filter
Culture Area 175 cm²

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.