Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface

Catalog No. p-200007435
Change view
Click to view available options
Culture Area:
175 cm²
25 cm²
75 cm²
Closure Type:
Filter
Solid
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Closure Type
12-565-666 25 cm² Filter
12-565-665 25 cm² Solid
12-565-664 75 cm² Filter
12-565-663 75 cm² Solid
12-565-662 175 cm² Filter
12-565-661 175 cm² Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-565-666 Supplier Thermo Scientific™ Supplier No. 156372
Add to Cart
Add to Cart

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Non-cytotoxic, Non-pyrogenic, USP Class VI
Description Nunc EasYFlask, 25 cm²
Material Polystyrene
For Use With (Application) General cell culture, cell therapy research, culture of fastidious adherent cells
Neck Style Angled
No. per Case 200 Flasks
No. per Pack 10 Flasks
Product Type Cell Culture Flask
Sterility Sterile
Class Class VI
Bottom Shape Flat
Closure Type Filter
Culture Area 25 cm²
Surface Treatment Nunclon Supra
Quantity 200/Cs.
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.