Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ IVF ICSI Dish

Catalog No. 12565023 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
51 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
12-565-023 51 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-565-023 Supplier Thermo Scientific™ Supplier No. 150265
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc™ IVF ICSI Dishes enable clear viewing of samples with a super-polished optical center and meet the exacting requirements of IVF testing, certification, and validation.

Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc IVF ICSI Dishes meet the exacting requirements of IVF testing, certification, and validation.

Innovative features for quality and ease-of-use

  • Maintain optimal temperature with true flat bottom
  • Low profile provides optimal angles during ICSI process
  • Clearly see sample with super polished optical center
  • Increased safety with easy grip design
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF

  • CE-marked (Notified Body 2797), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Certificates of conformity for your assurance on every lot

Sterile (SAL 10-6)

  • Non-pyrogenic (LAL test)
  • USP Class VI test
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE-marked, FDA-cleared, MEA-tested
Description IVF ICSI Dish, Lid w/ Airvent
Bottom Shape Flat
Color Clear
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Untreated
Type Dish
Well Count 1
No. per Case 120
No. per Pack 1
Height (Metric) 9 mm
Diameter (Metric) 51 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.