Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ IVF Dishes
GSA_VA

Catalog No. 12566353 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
35 mm
54 mm
88 mm
Height (Metric):
11 mm
14 mm
15 mm
Culture Area:
21.5 cm2
56.7 cm2
8.8 cm2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Height (Metric) Culture Area
12-566-353 54 mm 14 mm 21.5 cm2
12-566-352 35 mm 11 mm 8.8 cm2
12-566-351 88 mm 15 mm 56.7 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-566-353 Supplier Thermo Scientific™ Supplier No. 150270
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Product features for reliable results:

  • Facilitate gas exchange with vented lid
  • Available in 35mm, 60mm and 90mm sizes
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
Description IVF Dish, 60mm, Lid w/Airvent
Culture Area 21.5 cm2
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Non-treated
No. per Case 400
No. per Pack 10
Height (Metric) 14 mm
Diameter (Metric) 54 mm

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.