Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System

Catalog No. 125651 Shop All Thermo Scientific Products
Change view
Click to view available options
Wells:
1
2
4
8
Material:
CC2 Glass (mimics polylysine)
Description:
1-well Chamber Slide w/ removable well
2-well Chamber Slide w/ removable wells
4-well Chamber Slide w/ removable wells
8-well Chamber Slide w/ removable wells
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Material Description
12-565-1 8 CC2 Glass (mimics polylysine) 8-well Chamber Slide w/ removable wells
12-565-4 1 CC2 Glass (mimics polylysine) 1-well Chamber Slide w/ removable well
12-565-3 2 CC2 Glass (mimics polylysine) 2-well Chamber Slide w/ removable wells
12-565-2 4 CC2 Glass (mimics polylysine) 4-well Chamber Slide w/ removable wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12-565-1 Supplier Thermo Scientific™ Supplier No. 154941
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enable cell growth directly on a glass microscope slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System.

Enable cell growth (e.g. neurons) directly on an aminated glass slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System, which features a chemically modified CC2 growth surface that mimics the characters of polylysine coating. Available in 1-, 2-, 4-, and 8-well formats with removable media chambers.

  • Remains stable and consistent for over one year without refrigeration
  • Tabbed cover allows gas exchange while minimizing evaporation
  • Sterile
  • Removal tool separates chamber from glass
  • Provides binding sites optimal for fastidious cells
  • Light blue frosted writing area

Includes: Removal tool

TRUSTED_SUSTAINABILITY

Specifications

Wells 8
Culture Area 0.7 cm2
Lid Material Polystyrene
Description 8-well Chamber Slide w/ removable wells
Surface Treatment Cell Culture
Material CC2 Glass (mimics polylysine)
Volume (Metric) Working 0.2 to 0.5 mL
Includes Clear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 74 x 25 mm
Sterility Sterile
Closure Color Clear
Show More Show Less
What cell density should I use for Lab-Tek Chambered Slides and Lab-Tek II Chambered Slides?

Unfortunately, we do not have a recommended cell density for our LabTek Chamber Slides. However, you can refer to the following link, which provides recommended cell densities for our other cell culture products:
https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-protocols/cell-culture-useful-numbers.html

What stains and fixatives are compatible with Lab-Tek chambered slides and coverglasses?

Lab-Tek products are compatible with all common fixatives, including paraformaldehyde. Acetone at 100% and some mixtures of acetone and alcohols are not compatible with the polystyrene upper well structure and will result in clouding or other effects of chemical attack. We encourage careful testing within planned operating conditions before working with high-value samples.

What factors must I consider when working with Lab-Tek and Lab-Tek II chambered coverglass products?

Lab-Tek chambered coverglasses are not accessories to Lab-Tek chamber slides, but another complete product for cell culture. The chambers are simply adhered to a borosilicate coverglass (to serve as the culture surface) rather than a soda lime glass or Permanox microscope slide. Most customers select this format for compatibility with confocal imaging systems that require a #1.0, approx. 120 micron (Lab-Tek) or #1.5, approx. 180 micron (Lab-Tek II) substrate. Because the coverglass is very thin and fragile, the chambered coverglasses (unlike chamber slides) cannot be disassembled.

What are the different options available in Lab-Tek II chamber slides?

Lab-Tek II chambered glass slide systems are available in our standard high-purity water washed culture treatment or our “CC2” surface modification. The latter is a proprietary passive treatment that resembles poly-D-lysine, often improving culture performance when working with fastidious cells such as neuronal cultures.

What are the different options available in Lab-Tek chamber slides?

Lab-Tek chambered slide systems are based either on a standard soda lime glass microscope slide, or a culture-treated Permanox plastic slide. The latter may serve as a better culture surface for certain types of cells that don't grow well on glass, but may be of limited use for certain types of fluorescence microscopy due to the endogenous autofluorescence of plastic.

What is the cell culture treatment performed on glass Lab-Tek slides?

The standard glass Lab-Tek slide cell culture treatment consists of a proprietary, multi-stage high-purity water washing process. Unlike our plastic products for adherent cell culture (see Nunclon Delta), there is no need for further modification of the surface because glass is already hydrophilic and therefore already suitable for culture of many adherent cell lines.

What are the differences between Lab-Tek and Lab-Tek II chamber slide systems?

The main difference is the way the upper chamber structure is attached to the slide. Lab-Tek slides use a silicone gasket that can be peeled away once the chambers are removed after culture. Lab-Tek II slides use a hydrophobic acrylic adhesive that remains on the slide after chamber removal but (due to its very low profile) will not interfere with coverslips. The chamber geometry is also slightly different: on Lab-Tek slides the walls lean inward slightly to limit media evaporation, whereas on Lab-Tek II slides they are completely vertical.


Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.