Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
GSA_VA

Catalog No. 1256570 Shop All Thermo Scientific Products
Change view
Click to view available options
Plate Compatibility:
384-well Plate
96-well Plate
All Microplates
Sterility:
Non-sterile
Sterile
Condensation Rings:
No
Yes
No. per Pack:
1
5
20
25
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility Sterility Condensation Rings No. per Pack
12-565-70 96-well Plate Sterile Yes 1
12-565-476 All Microplates Sterile No 20
12-565-478 All Microplates Non-sterile No 20
12-565-462 All Microplates Non-sterile No 20
12-565-748 All Microplates Non-sterile Yes 25
12-565-749 All Microplates Sterile Yes 25
12-565-69 96-well Plate Non-sterile Yes 5
12-565-603 384-well Plate Sterile No 1
12-565-604 384-well Plate Non-sterile No 20
12-565-477 96-well Plate Sterile Yes 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 12-565-70 Supplier Thermo Scientific™ Supplier No. 264122
  • $406.50
    $220.65 / Case of 50

    Save $185.85

    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Dimensions (L x W x H) 5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Material Polystyrene
Sterility Sterile
No. of Wells 96
Height (Metric) 7.9 mm
Length (English) 5 in.
Length (Metric) 127 mm
Condensation Rings Yes
Height (English) 0.3 in.
No. per Case 50
No. per Pack 1
Dimensions (L x W) 5 x 3.35 in. (127 x 85 mm)
Evaporation Barrier Yes
Width (English) 3.35 in.
Width (Metric) 85 mm
Product Type Plate Lid
Product Line Nunc
Adhesive None
Temperature Range (Metric) -20°C to +80°C
Well Shape All well shapes
Plate Compatibility 96-well Plate
Cut-off Corners Long Side
Show More Show Less
Which Nunc products can I use for high-throughput, plate-based DNA plasmid minipreps?

We recommend using the following Nunc products:

- 96-Well Polypropylene DeepWell Storage Plate, 2 mL (Cat. No. 278743) (sterile growth plate)
- Microplate Lid for 96 microwell plate (Cat. No. 264122) (to cover plate during growth)
- Polyolefin Acrylate Sealing Tape (Cat. No. 232701) (to cover plate during mixing)
- Fritted 96-Well Deep Well Filter Plate (Cat. No. 278011) (for lysate clarification)
- Fritted 96-Well Deep Well Filter Plate with Glass Fiber Filter (Cat. No. 278010) (for DNA binding)
- 96-Well Polypropylene DeepWell Storage Plate, 1.3 mL (Cat. No. 260252) (reception plate)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.