Learn More
Thermo Scientific™ 96 Well Plate with Lid, Collagen I Coated Surface, Pack of 5
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
- Aseptically Manufactured
Specifications
Specifications
| Description | 96F well plate, Collagen I, clear, with lid |
| Material | Polystyrene |
| Volume (Metric) Well | 400 μL |
| Type | Microplate |
| Quantity | 5/Pk., 20/Cs. |
| Color | Clear |
| Surface Treatment | Collagen Type I |
| Sterility | Sterile |
| No. of Wells | 96 |
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Show More |
Frequently Asked Questions (FAQs)
96 Well Plate, Collagen I Coated Surface, Pack of 5 (Cat. No. 152038) provides 5 Cases with each Case containing 20 individual Nunc MicroWell Collagen Type I-coated 96-well microplates.
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.