Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
GSA_VA
Catalog No. 12565363 Shop All Thermo Scientific Products
Change view
Click to view available options
No. per Case:
108/Cs.
160/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
12-565-363 160/Cs.
12-565-311 108/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-565-363 Supplier Thermo Scientific™ Supplier No. 260860
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Edge
Add to Cart
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
  • Made from clear polystyrene with excellent optical properties, ideal for optical measurements.
  • Non-treated plate with hydrophobic surface is suited for suspension cell cultures and a variety of biochemical assays.
  • Cell culture plate received the Thermo Scientific™ Nunclon™ Delta surface treatment that promotes cell adhesion and growth
  • Collagen I and Poly-D-Lysine coated plates are enhanced for finicky adherent cells and biomolecule attachment
  • Different flange designs are compatible with either manual or robotic operations
  • Customized bar coding is available
Warranty: 90 days
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid With Lid
Description 96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 10, Case of 160)
Array 8 x 12
Packaging 160 Cases
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 160/Cs.
No. per Pack 10 Pk.
Quantity 10 Pk.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.